missing translation for 'onlineSavingsMsg'
Learn More

Germanium n-butoxide, 97+%

Catalog No. AA4081603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4081603 1 g
AA4081606 5 g
AA4081614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4081603 Supplier Thermo Scientific Chemicals Supplier No. 04081603

Oxide films | CAS: 25063-27-8

Germanium n-butoxide is used as oxide films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25063-27-8
MDL Number MFCD00015347
Synonym Tetra-n-butoxygermane

Specifications

Density 1.017 g/mL
Boiling Point 143°C (8 mmHg)
Flash Point 112°C (234°F)
Refractive Index 1.4255
Linear Formula Ge[O(CH2)3CH3]4
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 365.05
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Germanium n-butoxide
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 246-595-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only