Learn More
Germanium(IV) methoxide, 98%
CAS: 992-91-6 | C4H12GeO4
$323.05 - $1458.14
Chemical Identifiers
| CAS | 992-91-6 |
|---|---|
| Molecular Formula | C4H12GeO4 |
| MDL Number | MFCD00014881 |
| Synonym | tetramethoxygermanium, tetramethoxygermanium iv, methyl germanate iv |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1843706
|
Thermo Scientific Chemicals
A1843706 |
5 g |
Each for $323.05
|
|
|||||
|
AAA1843714
|
Thermo Scientific Chemicals
A1843714 |
25 g |
Each for $1,458.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 992-91-6 | |
| C4H12GeO4 | |
| MFCD00014881 | |
| tetramethoxygermanium, tetramethoxygermanium iv, methyl germanate iv |
Specifications
| 145°C | |
| 992-91-6 | |
| C4H12GeO4 | |
| MFCD00014881 | |
| 40°C (105°F) | |
| 196.73 | |
| Like Alcohol | |
| Moisture sensitive | |
| Germanium(IV) methoxide |
| -18°C | |
| 5 g | |
| (CH3O)4Ge | |
| UN2929 | |
| tetramethoxygermanium, tetramethoxygermanium iv, methyl germanate iv | |
| 98% | |
| 1.4015 | |
| 1.3254^2^0 |
Safety and Handling
GHS H Statement
H330-H226-H315-H319-H335
Fatal if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301+H311+H331-H315-H318-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only