missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Germanium(IV) methoxide, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 145°C |
| Melting Point | -18°C |
| Quantity | 5 g |
| Linear Formula | (CH3O)4Ge |
| UN Number | UN2929 |
| Flash Point | 40°C (105°F) |
| Formula Weight | 196.73 |
| Percent Purity | 98% |
| Odor | Like Alcohol |
| Refractive Index | 1.4015 |
| Show More |
Safety and Handling
| Hazard Category | H226-H301+H311+H331-H315-H318-H335 |
| Hazard Statement | GHS H Statement H330-H226-H315-H319-H335 Fatal if inhaled. Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: III |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only