Learn More
Germanium(IV) isopropoxide, 99.9% (metals basis)
CAS: 21154-48-3 | C12H28GeO4 | 308.982 g/mol
Supplier: Thermo Scientific Chemicals 04267306
Description
Germanium(IV) isopropoxide is used as precursor to prepare GeO2 organic silanes, which is used to study the application of optical wave guide. It is used in the synthesis of heterotactic polyactide and also involved in the macromolecule mediated synthesis of amorphous Germania.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Germanium (IV) isopropoxide | |
| Colorless | |
| 109°C (30 mmHg) | |
| Alcohol-like | |
| C12H28GeO4 | |
| Ge[OCH(CH3)2]4 | |
| 5 g | |
| germanium iv isopropoxide, tetraisopropoxygermane, germanium tetrapropan-2-olate, tetraisopropoxy-germane, tetraisopropoxygermanium, isopropyl germanate iv, germanium tetraisopropoxide | |
| CC(C)O[Ge](OC(C)C)(OC(C)C)OC(C)C | |
| 308.982 | |
| 308.94 | |
| Liquid |
| 21154-48-3 | |
| 1.025 g/mL | |
| 68°C (154°F) | |
| (metals basis) | |
| 1.414 | |
| MFCD00015339 | |
| Moisture sensitive | |
| PKLMYPSYVKAPOX-UHFFFAOYSA-N | |
| tetra(propan-2-yloxy)germane | |
| 16685043 | |
| 99.9% |
Chemical Identifiers
| 21154-48-3 | |
| 308.982 | |
| PKLMYPSYVKAPOX-UHFFFAOYSA-N | |
| 16685043 | |
| CC(C)O[Ge](OC(C)C)(OC(C)C)OC(C)C |
| C12H28GeO4 | |
| MFCD00015339 | |
| germanium iv isopropoxide, tetraisopropoxygermane, germanium tetrapropan-2-olate, tetraisopropoxy-germane, tetraisopropoxygermanium, isopropyl germanate iv, germanium tetraisopropoxide | |
| tetra(propan-2-yloxy)germane |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only