missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) iodide, 99.999% (metals basis)

Catalog No. p-7049326
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1400909 10 g
AA1400918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1400909 Supplier Thermo Scientific Chemicals Supplier No. 01400909

CAS: 13450-95-8 | GeI4 | 580.248 g/mol

Germanium(IV) iodide is a biochemical for proteomics research. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13450-95-8
Molecular Formula GeI4
Molecular Weight (g/mol) 580.248
MDL Number MFCD00016120
InChI Key CUDGTZJYMWAJFV-UHFFFAOYSA-N
Synonym germanium tetraiodide, germane, tetraiodo, germanium iv iodide, germanium iodide, acmc-1bq9p, germanium iv iodide trace metals basis
PubChem CID 83479
IUPAC Name tetraiodogermane
SMILES [Ge](I)(I)(I)I

Specifications

Melting Point 146°C
Physical Form Crystalline
Quantity 10 g
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 580.21
Percent Purity 99.999%
Sensitivity Moisture sensitive
Density 4.32 g/mL
Assay (metals basis)
Chemical Name or Material Germanium(IV) iodide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 236-613-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only