Learn More
Germanium ingot, zone refined, 99.9999% (metals basis), electrical resistivity: 40 ohm-cm
Form: Electrical Resistivity: 40 ohm-cm | CAS: 7440-56-4 | Ge | 72.63 g/mol
Supplier: Thermo Scientific Chemicals 01019279
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Germanium | |
| 7440-56-4 | |
| 120 g | |
| Ge | |
| element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder | |
| [Ge] | |
| 72.63 | |
| CHEBI:30441 | |
| Silver-colored ingot |
| Silver | |
| Ingot | |
| 99.9999% (Metals basis, 95% N type minimum) | |
| MFCD00085310 | |
| GNPVGFCGXDBREM-UHFFFAOYSA-N | |
| germanium | |
| 6326954 | |
| Odorless |
Chemical Identifiers
| 7440-56-4 | |
| 72.63 | |
| GNPVGFCGXDBREM-UHFFFAOYSA-N | |
| 6326954 | |
| germanium |
| Ge | |
| MFCD00085310 | |
| element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder | |
| CHEBI:30441 | |
| [Ge] |
Safety and Handling
EINECSNumber : 231-164-3
RTECSNumber : LY5200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only