missing translation for 'onlineSavingsMsg'
Learn More

Germanium(II) iodide, 99.99% (metals basis)

CAS: 13573-08-5 | GeI2 | 326.439 g/mol

$421.90 - $1754.71

Chemical Identifiers

CAS 13573-08-5
Molecular Formula GeI2
Molecular Weight (g/mol) 326.439
MDL Number MFCD00135537
InChI Key IAGYEMVJHPEPGE-UHFFFAOYSA-N
Synonym germanium diiodide, germanium ii iodide, germanium iodide, diiodogermanylidene, acmc-1c83a, germanium ii iodide trace metals basis
PubChem CID 6327215
IUPAC Name diiodogermanium
SMILES [Ge](I)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1401303
View Documents
Thermo Scientific Chemicals
01401303
1 g
Each for $421.90
Only null left
 
AA1401306
View Documents
Thermo Scientific Chemicals
01401306
5 g
Each for $1,754.71
Only null left
 
Description

Description

Germanium(II) iodide acts as a reagent that forms stable ionic adducts with carbenes. It is also used in the preparation of allylgermanes by reacting with allylstannanes. Further, it reacts with allyl bromide in the presence of zinc iodide to get allylgermanium(IV), which on reacts with carbonyl compounds to give the corresponding homoallylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13573-08-5
326.439
IAGYEMVJHPEPGE-UHFFFAOYSA-N
6327215
[Ge](I)I
GeI2
MFCD00135537
germanium diiodide, germanium ii iodide, germanium iodide, diiodogermanylidene, acmc-1c83a, germanium ii iodide trace metals basis
diiodogermanium

Specifications

13573-08-5
1 g
MFCD00135537
germanium diiodide, germanium ii iodide, germanium iodide, diiodogermanylidene, acmc-1c83a, germanium ii iodide trace metals basis
IAGYEMVJHPEPGE-UHFFFAOYSA-N
diiodogermanium
6327215
99.99%
5.37 g/mL
(metals basis)
Yellow to Brown
GeI2
UN3260
Soluble in dilute hydrochloric acid.
[Ge](I)I
326.439
326.4
Air and moisture sensitive
Germanium(II) iodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 236-998-1

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only