missing translation for 'onlineSavingsMsg'
Learn More

Germanium, 99.999%, (trace metal basis), powder

CAS: 7440-56-4 | Ge | 72.63 g/mol

$441.44 - $441.44

Chemical Identifiers

CAS 7440-56-4
Molecular Formula Ge
Molecular Weight (g/mol) 72.63
MDL Number MFCD00085310
InChI Key GNPVGFCGXDBREM-UHFFFAOYSA-N
Synonym element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
PubChem CID 6326954
ChEBI CHEBI:30441
IUPAC Name germanium
SMILES [Ge]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC193110100
SDS View Documents
Thermo Scientific Chemicals
193110100
10 g Glass bottle
Each for $441.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

7440-56-4
72.63
GNPVGFCGXDBREM-UHFFFAOYSA-N
6326954
germanium
Ge
MFCD00085310
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
CHEBI:30441
[Ge]

Specifications

2830.0°C
7440-56-4
Glistening Crystalline Powder
99.999%
MFCD00085310
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
GNPVGFCGXDBREM-UHFFFAOYSA-N
germanium
6326954
72.59
Trace Metal Basis
5.35
Germanium, 99.999%
937.0°C
Black to Gray
10 g
Ge
15, 4441
Solubility in water: insoluble. Other solubilities: insoluble in hcl and diluted alkali hydroxides
[Ge]
72.63
CHEBI:30441
99.999%
Glass bottle
5.3500g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-164-3

RTECSNumber : LY5200000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only