missing translation for 'onlineSavingsMsg'
Learn More

Geranyl Nitrile 95.0+%, TCI America™

Numéro de catalogue. G024125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
G024125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. G024125ML Fournisseur TCI America Code fournisseur G024125ML
Il en reste null

Identifiants chimiques

CAS 5146-66-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00001926
InChI Key HLCSDJLATUNSSI-JXMROGBWSA-N
Synonym geranyl nitrile, citralva, 3,7-dimethylocta-2,6-dienenitrile, geranonitrile, 3,7-dimethyl-2,6-octadienenitrile, e-3,7-dimethyl-2,6-octadienenitrile, unii-w889cv0e9f, 2e-3,7-dimethylocta-2,6-dienenitrile, 2,6-octadienenitrile, 3,7-dimethyl-, e, 2-geranonitrile
PubChem CID 1551246
IUPAC Name (2E)-3,7-dimethylocta-2,6-dienenitrile
SMILES CC(=CCCC(=CC#N)C)C

Spécifications

Color Yellow
Boiling Point 222°C
Quantity 25 mL
Formula Weight 149.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Geranyl Nitrile [mixture of (E)- and (Z)- isomers, (1:1)]
EINECSNumber (2)-1515
RTECSNumber RG5370000
TSCA Yes