missing translation for 'onlineSavingsMsg'
Learn More

Geranyl-linalool (mixture of isomers) 90.0+%, TCI America™

Catalog No. g022125ml
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G022125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G022125ML Supplier TCI America Supplier No. G022125ML
Only null left

Chemical Identifiers

CAS 68931-30-6
Molecular Formula C20H34O
Molecular Weight (g/mol) 290.491
MDL Number MFCD00059363
InChI Key IQDXAJNQKSIPGB-HQSZAHFGSA-N
Synonym 3,7,11,15-Tetramethyl-1,6,10,14-hexadecatetraen-3-ol
PubChem CID 5365872
ChEBI CHEBI:74299
IUPAC Name (6E,10E)-3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol
SMILES CC(=CCCC(=CCCC(=CCCC(C)(C=C)O)C)C)C

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 mL
Formula Weight 290.49
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Geranyl-linalool (mixture of isomers)
RTECSNumber MM0335000
TSCA Yes