missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Geranyl Formate 55.0+%, TCI America™
Supplier: TCI America G021825ML
| Quantity | 25 mL |
|---|
Chemical Identifiers
| 105-86-2 | |
| 182.263 | |
| FQMZVFJYMPNUCT-YRNVUSSQSA-N | |
| CHEBI:31648 | |
| CC(=CCCC(=CCOC=O)C)C |
| C11H18O2 | |
| MFCD00021047 | |
| 5282109 | |
| [(2E)-3,7-dimethylocta-2,6-dienyl] formate |
Specifications
| Geranyl Formate [for Perfumery] | |
| Colorless | |
| C11H18O2 | |
| MFCD00021047 | |
| CC(=CCCC(=CCOC=O)C)C | |
| 182.263 | |
| CHEBI:31648 | |
| ≥55.0% (GC) |
| 105-86-2 | |
| 92°C | |
| 25 mL | |
| FQMZVFJYMPNUCT-YRNVUSSQSA-N | |
| [(2E)-3,7-dimethylocta-2,6-dienyl] formate | |
| 5282109 | |
| 182.26 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2529
RTECSNumber : RG5925700
TSCA : Yes