missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Geranyl Acetate 70.0+%, TCI America™
$39.87 - $307.57
Chemical Identifiers
| CAS | 105-87-3 |
|---|---|
| Molecular Formula | C12H20O2 |
| Molecular Weight (g/mol) | 196.29 |
| MDL Number | MFCD00015037 |
| InChI Key | HIGQPQRQIQDZMP-DHZHZOJOSA-N |
| Synonym | geranyl acetate, geraniol acetate, bay pine oyster oil, acetic acid, geraniol ester, trans-3,7-dimethyl-2,6-octadien-1-yl acetate, geranyl ethanoate, acetic acid, geranyl ester, unii-3w81yg7p9r, geranyl acetate, cis, 3,7-dimethyl-2,6-octadienyl acetate |
| PubChem CID | 1549026 |
| ChEBI | CHEBI:5331 |
| IUPAC Name | (2E)-3,7-dimethylocta-2,6-dien-1-yl acetate |
| SMILES | CC(C)=CCC\C(C)=C\COC(C)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
G002825ML
|
TCI America
G002825ML |
25 mL |
Each for $39.87
|
|
|||||
|
G0028100ML
|
TCI America
G0028100ML |
100 mL |
Each for $102.52
|
|
|||||
|
G0028500ML
|
TCI America
G0028500ML |
500 mL |
Each for $307.57
|
|
|||||
Chemical Identifiers
| 105-87-3 | |
| 196.29 | |
| HIGQPQRQIQDZMP-DHZHZOJOSA-N | |
| 1549026 | |
| (2E)-3,7-dimethylocta-2,6-dien-1-yl acetate |
| C12H20O2 | |
| MFCD00015037 | |
| geranyl acetate, geraniol acetate, bay pine oyster oil, acetic acid, geraniol ester, trans-3,7-dimethyl-2,6-octadien-1-yl acetate, geranyl ethanoate, acetic acid, geranyl ester, unii-3w81yg7p9r, geranyl acetate, cis, 3,7-dimethyl-2,6-octadienyl acetate | |
| CHEBI:5331 | |
| CC(C)=CCC\C(C)=C\COC(C)=O |
Specifications
| 105-87-3 | |
| 120°C | |
| MFCD00015037 | |
| geranyl acetate, geraniol acetate, bay pine oyster oil, acetic acid, geraniol ester, trans-3,7-dimethyl-2,6-octadien-1-yl acetate, geranyl ethanoate, acetic acid, geranyl ester, unii-3w81yg7p9r, geranyl acetate, cis, 3,7-dimethyl-2,6-octadienyl acetate | |
| CC(C)=CCC\C(C)=C\COC(C)=O | |
| 196.29 | |
| CHEBI:5331 | |
| ≥70.0% (GC) | |
| Geranyl Acetate |
| Colorless | |
| C12H20O2 | |
| 25 mL | |
| HIGQPQRQIQDZMP-DHZHZOJOSA-N | |
| (2E)-3,7-dimethylocta-2,6-dien-1-yl acetate | |
| 1549026 | |
| 196.29 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2535
RTECSNumber : RG5920000
TSCA : Yes