missing translation for 'onlineSavingsMsg'
Learn More

Geranyl Acetate 70.0+%, TCI America™

Catalog No. G0028500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0028500ML 500 mL
G002825ML 25 mL
G0028100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G0028500ML Supplier TCI America Supplier No. G0028500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105-87-3
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00015037
InChI Key HIGQPQRQIQDZMP-DHZHZOJOSA-N
Synonym geranyl acetate, geraniol acetate, bay pine oyster oil, acetic acid, geraniol ester, trans-3,7-dimethyl-2,6-octadien-1-yl acetate, geranyl ethanoate, acetic acid, geranyl ester, unii-3w81yg7p9r, geranyl acetate, cis, 3,7-dimethyl-2,6-octadienyl acetate
PubChem CID 1549026
ChEBI CHEBI:5331
IUPAC Name (2E)-3,7-dimethylocta-2,6-dien-1-yl acetate
SMILES CC(C)=CCC\C(C)=C\COC(C)=O

Specifications

Color Colorless
Boiling Point 120°C
Quantity 500 mL
Formula Weight 196.29
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material Geranyl Acetate
EINECSNumber (2)-2535
RTECSNumber RG5920000
TSCA Yes