missing translation for 'onlineSavingsMsg'
Learn More

Geranyl acetate, 98%

Catalog No. AAA1986414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1986414 25 g
AAA1986422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1986414 Supplier Thermo Scientific Chemicals Supplier No. A1986414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 105-87-3 | C12H20O2 | 196.29 g/mol

It is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient. It is used particularly in rose, lavender and geranium formulations where a sweet fruity or citrus aroma is desired. It is found to have has anti-oxidant, anti-inflammatory and anti-proliferative activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient. It is used particularly in rose, lavender and geranium formulations where a sweet fruity or citrus aroma is desired. It is found to have has anti-oxidant, anti-inflammatory and anti-proliferative activities.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 105-87-3
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00015037
InChI Key HIGQPQRQIQDZMP-DHZHZOJOSA-N
Synonym geranyl acetate, geraniol acetate, bay pine oyster oil, acetic acid, geraniol ester, trans-3,7-dimethyl-2,6-octadien-1-yl acetate, geranyl ethanoate, acetic acid, geranyl ester, unii-3w81yg7p9r, geranyl acetate, cis, 3,7-dimethyl-2,6-octadienyl acetate
PubChem CID 1549026
ChEBI CHEBI:5331
IUPAC Name [(2E)-3,7-dimethylocta-2,6-dienyl] acetate
SMILES CC(C)=CCC\C(C)=C\COC(C)=O

Specifications

Density 0.91
Boiling Point 137°C to 139°C (25 mmHg)
Flash Point 104°C (219°F)
Odor Pleasant
Assay Percent Range 0.98
Refractive Index 1.461
Quantity 25 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 196.29
Percent Purity 98%
Chemical Name or Material Geranyl acetate
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 203-341-5
RTECSNumber RG5920000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only