missing translation for 'onlineSavingsMsg'
Learn More

Geraniol, 99%

Catalog No. AC410901000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410901000 100 g Glass bottle
AC410900050 5 g Glass bottle
AC410900250 25 g Glass bottle
AC410905000 500 g Glass bottle
AC410900025 2.5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC410901000 Supplier Thermo Scientific Chemicals Supplier No. 410901000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-24-1 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-24-1
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00002917
InChI Key GLZPCOQZEFWAFX-JXMROGBWSA-N
Synonym geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
PubChem CID 637566
ChEBI CHEBI:17447
IUPAC Name (2E)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(=CCCC(=CCO)C)C

Specifications

Melting Point -15.0°C
Color Colorless to Yellow
Density 0.8780g/mL
Boiling Point 229.0°C to 230.0°C (757.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
Refractive Index 1.4750 to 1.4790
Quantity 100 g
Beilstein 01,457
Merck Index 15,4437
Specific Gravity 0.878
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with alcohol and ether, ethanol 50 vol. % (20°C):1:10 v:v
Formula Weight 154.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Geraniol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 203-377-1

RUO – Research Use Only