missing translation for 'onlineSavingsMsg'
Learn More

Geraniol, 99%

CAS: 106-24-1 | C10H18O | 154.25 g/mol

Supplier:  Thermo Scientific Chemicals 410900250

Quantity 25 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC410900250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-24-1
154.25
GLZPCOQZEFWAFX-JXMROGBWSA-N
637566
(2E)-3,7-dimethylocta-2,6-dien-1-ol
C10H18O
MFCD00002917
geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
CHEBI:17447
CC(=CCCC(=CCO)C)C

Specifications

Geraniol
-15.0°C
0.8780g/mL
76°C
98.5% min. (GC)
C10H18O
1.4750 to 1.4790
MFCD00002917
15,4437
geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
GLZPCOQZEFWAFX-JXMROGBWSA-N
(2E)-3,7-dimethylocta-2,6-dien-1-ol
637566
154.25
Liquid
106-24-1
Colorless to Yellow
229.0°C to 230.0°C (757.0 mmHg)
Authentic
Glass bottle
(CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
25 g
01,457
0.878
Solubility in water: practically insoluble. Other solubilities: miscible with alcohol and ether, ethanol 50 vol. % (20°C):1:10 v:v
CC(=CCCC(=CCO)C)C
154.25
CHEBI:17447
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 203-377-1

SDS
Documents

Documents

Promotions

Promotions