missing translation for 'onlineSavingsMsg'
Learn More

Geraniol, 97%

CAS: 106-24-1 | C10H18O | 154.253 g/mol

Supplier:  Thermo Scientific Chemicals A137360B

Quantity 1000 g

 View more versions of this product

Catalog No. AAA137360B


Only null left

Description

Description

Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.

Solubility
Not miscible in water.

Notes
Incompatible with Strong oxidizing agents, acid chlorides, acid anhydrides. Keep away from heat. Air sensitive.
Specifications

Chemical Identifiers

106-24-1
154.253
GLZPCOQZEFWAFX-JXMROGBWSA-N
637566
(2E)-3,7-dimethylocta-2,6-dien-1-ol
C10H18O
MFCD00002917
geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
CHEBI:17447
CC(=CCCC(=CCO)C)C

Specifications

Geraniol
-15°C
114°C to 115°C (12 mmHg)
Floral
1.476
MFCD00002917
14,4403
Not miscible in water.
CC(=CCCC(=CCO)C)C
154.253
CHEBI:17447
97%
106-24-1
0.884
101°C (213°F)
C10H18O
1000 g
1722456
geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
GLZPCOQZEFWAFX-JXMROGBWSA-N
(2E)-3,7-dimethylocta-2,6-dien-1-ol
637566
154.25
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P370+P378q-P501c

H227-H315-H317-H318

EINECSNumber : 203-377-1

RTECSNumber : RG5830000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions