missing translation for 'onlineSavingsMsg'
Learn More

Geranic acid, 98% (sum of isomers)

CAS: 459-80-3 | C10H16O2 | 168.24 g/mol

$133.18 - $365.60

Chemical Identifiers

CAS 459-80-3
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00036493,MFCD20542837
InChI Key ZHYZQXUYZJNEHD-VQHVLOKHSA-N
Synonym geranic acid, 3,7-dimethylocta-2,6-dienoic acid, decaprenoic acid, trans-geranic acid, 3,7-dimethyl-2,6-octadienoic acid, 3,7-dimethyl-2e,6-octadienoic acid, neric acid, 2e-3,7-dimethylocta-2,6-dienoic acid, e-3,7-dimethylocta-2,6-dienoic acid, nerolic acid
PubChem CID 5275520
ChEBI CHEBI:67264
IUPAC Name (2E)-3,7-dimethylocta-2,6-dienoic acid
SMILES CC(C)=CCC\C(C)=C\C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1875014
View Documents
Thermo Scientific Chemicals
A1875014
25 g
Each for $133.18
Only null left
 
AAA1875022
View Documents
Thermo Scientific Chemicals
A1875022
100 g
Each for $365.60
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

459-80-3
168.24
ZHYZQXUYZJNEHD-VQHVLOKHSA-N
5275520
(2E)-3,7-dimethylocta-2,6-dienoic acid
C10H16O2
MFCD00036493,MFCD20542837
geranic acid, 3,7-dimethylocta-2,6-dienoic acid, decaprenoic acid, trans-geranic acid, 3,7-dimethyl-2,6-octadienoic acid, 3,7-dimethyl-2e,6-octadienoic acid, neric acid, 2e-3,7-dimethylocta-2,6-dienoic acid, e-3,7-dimethylocta-2,6-dienoic acid, nerolic acid
CHEBI:67264
CC(C)=CCC\C(C)=C\C(O)=O

Specifications

459-80-3
249°C to 251°C
C10H16O2
(CH3)2C=CHCH2CH2C(CH3)=CHCO2H
25 g
geranic acid, 3,7-dimethylocta-2,6-dienoic acid, decaprenoic acid, trans-geranic acid, 3,7-dimethyl-2,6-octadienoic acid, 3,7-dimethyl-2e,6-octadienoic acid, neric acid, 2e-3,7-dimethylocta-2,6-dienoic acid, e-3,7-dimethylocta-2,6-dienoic acid, nerolic acid
CC(C)=CCC\C(C)=C\C(O)=O
168.24
CHEBI:67264
≥90%
0.955
133°C (271°F)
1.484
MFCD00036493,MFCD20542837
1763804
ZHYZQXUYZJNEHD-VQHVLOKHSA-N
(2E)-3,7-dimethylocta-2,6-dienoic acid
5275520
168.24
Geranic acid, sum of isomers
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H312-H315-H319-H335

EINECSNumber : 207-299-9

RTECSNumber : RG5380000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only