missing translation for 'onlineSavingsMsg'
Learn More

Geranic acid, 98% (sum of isomers)

Catalog No. AAA1875014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1875014 25 g
AAA1875022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1875014 Supplier Thermo Scientific Chemicals Supplier No. A1875014

CAS: 459-80-3 | C10H16O2 | 168.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 459-80-3
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00036493,MFCD20542837
InChI Key ZHYZQXUYZJNEHD-VQHVLOKHSA-N
Synonym geranic acid, 3,7-dimethylocta-2,6-dienoic acid, decaprenoic acid, trans-geranic acid, 3,7-dimethyl-2,6-octadienoic acid, 3,7-dimethyl-2e,6-octadienoic acid, neric acid, 2e-3,7-dimethylocta-2,6-dienoic acid, e-3,7-dimethylocta-2,6-dienoic acid, nerolic acid
PubChem CID 5275520
ChEBI CHEBI:67264
IUPAC Name (2E)-3,7-dimethylocta-2,6-dienoic acid
SMILES CC(C)=CCC\C(C)=C\C(O)=O

Specifications

Density 0.955
Boiling Point 249°C to 251°C
Flash Point 133°C (271°F)
Refractive Index 1.484
Linear Formula (CH3)2C=CHCH2CH2C(CH3)=CHCO2H
Quantity 25 g
Beilstein 1763804
Formula Weight 168.24
Percent Purity ≥90%
Chemical Name or Material Geranic acid, sum of isomers
Hazard Category H312-H315-H319-H335
Hazard Statement GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-299-9
RTECSNumber RG5380000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only