missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Geometrical Models

Catalog No. S12913
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S12913 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12913 Supplier Eisco™ Supplier No. PH0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains various geometrical models and figures.

  • Made of hard wood
  • Consists of 16 pieces
  • Supplied in wooden box

Specifications

Grade K to 3
Includes Contains various geometrical models and figures
Material Hardwood
Quantity 16 Pk.
Class Size 1 Class
Product Type Geometrical Model