missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GENTEC™ Genstar Technologies™ Gentec™ M22 Series Metering Valves
With maximum operating pressure of 1000 psi (69 bar)
Supplier: GENTEC™ SS MV 22 TF4
Includes: Handle, Panel Nut, Bonnet Sleeve, Valve Stem, Bonnet, Lower Gland, O-ring, Body
Description
- GENLOK and NPT end connections
- Panel mounting available
- Cleaned and degreased for oxygen service according to CGA G-4.1 and ASTM G93 Class C, respectively
Chemical Identifiers
| Valve | |
| -23°C to +204°C | |
| 316 Stainless Steel (Handle, Panel Nut, Bonnet Sleeve, Valve Stem, Bonnet, Body), PTFE (Lower Gland), Fluorine Rubber (O-Ring) |
| -10°F to +400°F | |
| Handle, Panel Nut, Bonnet Sleeve, Valve Stem, Bonnet, Lower Gland, O-ring Fluorine, Body | |
| 1000 psi |
Specifications
| Valve | |
| -23°C to +204°C | |
| Handle, Panel Nut, Bonnet Sleeve, Valve Stem, Bonnet, Lower Gland, O-ring Fluorine, Body | |
| 316 Stainless Steel (Handle, Panel Nut, Bonnet Sleeve, Valve Stem, Bonnet, Body), PTFE (Lower Gland), Fluorine Rubber (O-Ring) |
| -10°F to +400°F | |
| SS-MV22-TF4 | |
| 1/4 in. GENLOK End Connections | |
| 1000 psi |