Learn More
Amsbio LLC (formerly Genlantis) TurboCells™ BL21Competent Cells
Catalog No. C303020
Encompass
Change view
Click to view available options
Product Type:
BL21 (DE3) Competent E. coli
BL21(DE3) pLysS Competent E. coli
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
C303020 BL21(DE3) pLysS Competent E. coli
C302020 BL21 (DE3) Competent E. coli
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C303020 Supplier Amsbio LLC (formerly Genlantis) Supplier No. C303020
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
High-level E. coli expression using three-minute transformation protocol
  • Achieve high-level recombinant protein expression with the T7 expression system
  • No hour-long recovery step or inconvenient 42°C water bath
  • Convenient and efficient single-use packaging
  • Two useful genotypes
Specifications
For Use With (Application) High-level E. coli expression using 3 min. transformation protocol
Product Type BL21(DE3) pLysS Competent E. coli
Quantity 20 x 50 μL