missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ PrimaPure™ Preadipocytes

Catalog No. PH80205A
Encompass
Change view
Click to view available options
Cell Type:
HPAd, adult heart
HPAd, adult subcutaneous
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type
PH80205A HPAd, adult heart
PH80305A HPAd, adult subcutaneous
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PH80205A Supplier Genlantis™ Supplier No. PH80205A

High-quality human preadipocytes from adult heart or subcutaneous tissue

Each lot functionally characterized

Same quality standard as the PrimaPure human cell products

Each lot tested negative for HIV, Hepatitis B C, mycoplasma, bacteria, yeast, and fungi

Performance guaranteed with Genlantis media and reagents
Human Preadipocytes (HPAd) are fibroblast like precursor cells derived from human adipose tissue.

Specifications

Cell Type HPAd, adult heart
For Use With (Application) Study insulin-stimulated glucose transport, growth hormone enhanced lypolysis and obese gene expression
Media Recommendation Human Preadipocyte Growth Medium
Quantity >500000 Cells
Source Human Adipose Tissue
Sufficient For 16 Population Doublings