missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ PrimaPure™ Human Umbilical Vein Endothelial Cells (HUVEC) and Kits

Catalog No. PH20005NP
Encompass
Change view
Click to view available options
Quantity:
Pooled, >500000 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PH20005NP Pooled, >500000 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PH20005NP Supplier Genlantis™ Supplier No. PH20005NP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Functionally characterized

Human Umbilical Vein Endothelial Cells (HUVEC) are isolated from normal human umbilical vein. They are cryopreserved at the end of primary culture and can be cultured and propagated at least 16 population doublings. HUVEC are responsive to cytokine stimulation in the expression of cell adhesion molecules. These cell systems are commonly used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation and fibrinolysis.

  • Single donors to allow variability comparisons
  • Each lot tested negative for HIV, Hepatitis B and C, mycoplasma, bacteria, yeast, and fungi
  • Performance guaranteed with Genlantis media and reagents

Specifications

Cell Type Human Umbilical Vein Endothelial Cells (HUVEC, Neonatal)
For Use With (Application) Used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation, fibrinolysis
Media Recommendation Endothelial Cell Growth Medium
Quantity Pooled, >500000 Cells
Source Human Umbilical Vein
Sufficient For 16 Population Doublings