missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ PrimaPure™ Human Umbilical Vein Endothelial Cells (HUVEC) and Kits

Catalog No. PH20005N
Encompass
Change view
Click to view available options
Quantity:
>500000 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PH20005N >500000 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PH20005N Supplier Genlantis™ Supplier No. PH20005N

missing translation for 'validMainframeMsg'

Specifications

Cell Type Human Umbilical Vein Endothelial Cells (HUVEC, Neonatal)
For Use With (Application) Used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation, fibrinolysis
Media Recommendation Endothelial Cell Growth Medium
Quantity >500000 Cells
Source Human Umbilical Vein
Sufficient For 16 Population Doublings