missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ PrimaPure™ Fibroblasts Kits

Catalog No. PH10605NK
Encompass
Change view
Click to view available options
Cell Type:
Adult HDF
Fetal HDF
Neonatal HDF
Source:
Human Adult Skin
Human Fetal
Human Neonatal Foreskin
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type Source
PH10605NK Neonatal HDF Human Neonatal Foreskin
PH10605AK Adult HDF Human Adult Skin
PH10605FK Fetal HDF Human Fetal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PH10605NK Supplier Genlantis™ Supplier No. PH10605NK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-quality human adult, fetal, and neonatal fibroblasts

Specifications

Content And Storage 4° and -20°C
Cell Type Neonatal HDF
For Use With (Application) For in-vitro analysis of fibroblast growth, migration and collagen metabolism in wound healing
Includes One ampoule of cryopreserved HDF, 500mL of HDF cell growth medium, one subculture reagent kit (100mL each of HEPES buffered saline solution, trypsin/EDTA, and trypsin neutralizing solution)
Media Recommendation Fibroblast Growth Medium
Quantity 1 Kit
Source Human Neonatal Foreskin
Sufficient For 16 Population Doublings