missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genlantis™ PrimaPure™ Fibroblasts Kits
Specifications
Specifications
| Content And Storage | 4° and -20°C |
| Cell Type | Fetal HDF |
| For Use With (Application) | For in-vitro analysis of fibroblast growth, migration and collagen metabolism in wound healing |
| Includes | One ampoule of cryopreserved HDF, 500mL of HDF cell growth medium, one subculture reagent kit (100mL each of HEPES buffered saline solution, trypsin/EDTA, and trypsin neutralizing solution) |
| Media Recommendation | Fibroblast Growth Medium |
| Quantity | 1 Kit |
| Source | Human Fetal |
| Sufficient For | 16 Population Doublings |