Learn More
Genlantis™ PrimaPure™ Epidermal Keratinocytes and Kits
High-quality primary adult, fetal, and neonatal HEK
Supplier:  Genlantis™ PH10205FK
Encompass
 View more versions of this product
Catalog No. PH10205FK
Only null left
Description
Description
  • Each lot is functionally characterized
  • Single donors to allow variability comparisons
  • Each lot tested negative for HIV, Hepatitis B C, mycoplasma, bacteria, yeast, and fungi
  • Supports 16 population doublings
Specifications
Chemical Identifiers
4° and -20°C
In-vitro model for dermal toxicology tests in screening new skin products and for developing dermal skin substitutes, for investigations of human epidermal development, differentiation, and cellular aspects of skin diseases
Human Neonatal Foreskin or Adult Skin
16 Population Doublings
Specifications
4° and -20°C
In-vitro model for dermal toxicology tests in screening new skin products and for developing dermal skin substitutes, for investigations of human epidermal development, differentiation, and cellular aspects of skin diseases
Keratinocyte Serum-free Growth Medium
Human Neonatal Foreskin or Adult Skin
Fetal HEK
One ampoule of cryopreserved HEK, 500mL of keratinocyte Serum-free growth medium, one subculture reagent kit (100mL each of HEPES buffered saline solution, trypsin/EDTA, and trypsin neutralizing solution)
1 Kit
16 Population Doublings
Product Suggestions
Product Suggestions