missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ PrimaPure™ Epidermal Keratinocytes and Kits

Catalog No. PH10205F
Encompass
Change view
Click to view available options
Cell Type:
Adult HEK
Fetal HEK
Neonatal HEK
Quantity:
>500000 Cells
1 Kit
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type Quantity
PH10205F Fetal HEK >500000 Cells
PH10205A Adult HEK >500000 Cells
PH10205AK Adult HEK 1 Kit
PH10205FK Fetal HEK 1 Kit
PH10205N Neonatal HEK >500000 Cells
PH10205NK Neonatal HEK 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. PH10205F Supplier Genlantis™ Supplier No. PH10205F
Only null left

High-quality primary adult, fetal, and neonatal HEK

  • Each lot is functionally characterized
  • Single donors to allow variability comparisons
  • Each lot tested negative for HIV, Hepatitis B C, mycoplasma, bacteria, yeast, and fungi
  • Supports 16 population doublings

Specifications

Content And Storage 4° and -20°C
Cell Type Fetal HEK
For Use With (Application) In-vitro model for dermal toxicology tests in screening new skin products and for developing dermal skin substitutes, for investigations of human epidermal development, differentiation, and cellular aspects of skin diseases
Includes
Media Recommendation Keratinocyte Serum-free Growth Medium
Quantity >500000 Cells
Source Human Neonatal Foreskin or Adult Skin
Sufficient For 16 Population Doublings