missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genlantis™ PrimaPure™ Epidermal Keratinocytes and Kits
Description
- Each lot is functionally characterized
- Single donors to allow variability comparisons
- Each lot tested negative for HIV, Hepatitis B C, mycoplasma, bacteria, yeast, and fungi
- Supports 16 population doublings
Specifications
Specifications
| Content And Storage | 4° and -20°C |
| Cell Type | Adult HEK |
| For Use With (Application) | In-vitro model for dermal toxicology tests in screening new skin products and for developing dermal skin substitutes, for investigations of human epidermal development, differentiation, and cellular aspects of skin diseases |
| Includes | One ampoule of cryopreserved HEK, 500mL of keratinocyte Serum-free growth medium, one subculture reagent kit (100mL each of HEPES buffered saline solution, trypsin/EDTA, and trypsin neutralizing solution) |
| Media Recommendation | Keratinocyte, Adult, Serum-free Growth Medium |
| Quantity | 1 Kit |
| Source | Human Neonatal Foreskin or Adult Skin |
| Sufficient For | 16 Population Doublings |