Learn More
Genlantis™ NeuroPORTER™ Transfection Reagent
Description
- High-level transfection efficiencies in neuronal cell lines, glial cells, and certain primary neurons
- Optimized protocols for primary neurons, neuronal cell lines, and glial cells
- Minimized cytotoxicity
- Unique cationic lipid specifically formulated for efficient DNA transfection into neuronal cell lines, glial cells, and certain primary neurons
Specifications
Specifications
| For Use With (Application) | Unique cationic Lipid specifically formulated for efficient DNA transfection into neuronal cell lines, glial cells and certain primary neurons |
| Includes | 1 Vial dried Lipid film, 1.5mL Hydration Buffer, 7.5mL DNA diluents |
| Quantity | 1.5 mL |