missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genlantis™ DNA Markers and Ladders
Fragment size from 11 base pairs to 15kb for a variety of electrophoretic applications
$377.42 - $484.17
Specifications
| Product Type | DNA Molecular Weight Marker/Ladder |
|---|---|
| For Use With (Application) | Determining the size of DNA fragment |
| Storage Requirements | −20°C |
Description
- High quality DNA molecular weight markers for a variety of electrophoretic applications
- Ideal for determining the size of a wide range of DNA fragments
- Sensitive and quick staining of DNA
- Available in various formulations to provide you with maximum versatility
Specifications
| DNA Molecular Weight Marker/Ladder | |
| Determining the size of DNA fragment | |
| −20°C |