missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genlantis™ Coomassie™ Stains
For rapid protein and DNA detection
$169.88 - $213.48
Specifications
| Color | Brilliant Blue |
|---|---|
| Product Type | Protein Stain |
| For Use With (Application) | Protein and DNA detection |
| Molecular Weight (g/mol) | 854.04 |
Description
- Coomassie™ Brilliant Blue G-250 is often used for detecting protein concentration by Bradford method
- Coomassie™ Brilliant Blue R-250 offers more sensitive detection than G-250
Specifications
| Brilliant Blue | |
| Protein Stain | |
| Protein and DNA detection | |
| 854.04 |