missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genlantis™ BioCooler™ Midi Cold Box
Convenient mid-sized benchtop cooler for all temperature sensitive reagents
Supplier: Genlantis™ E100500
Description
- Convenient capacity of 12 vials (1.5 to 2.0mL each)
- Temperatures of -14°C or less up to 90 minutes on the benchtop
- Small benchtop footprint
- Rugged high impact resistant ABS plastic construction
- Hinged clear polycarbonate lid with safety latches to prevents spills but allows visualization of samples
Specifications
| Polycarbonate Hinged | |
| 0.85 kg | |
| BioCooler Mini Cold Box | |
| 12 (1.5 or 2mL at -14°C or less up to 90 min.) |
| 1.87 lb. | |
| ABS | |
| White | |
| -14°C |