missing translation for 'onlineSavingsMsg'
Learn More

Geniposide 95.0+%, TCI America™

Catalog No. G0385100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0385100MG 100 mg
G03851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0385100MG Supplier TCI America Supplier No. G0385100MG

Chemical Identifiers

CAS 24512-63-8
Molecular Formula C17H24O10
Molecular Weight (g/mol) 388.369
MDL Number MFCD00016659
InChI Key IBFYXTRXDNAPMM-YFAICUPXSA-N
Synonym Methyl (1S)-1alpha-(beta-D-Glucopyranosyloxy)-7-(hydroxymethyl)-1,4aalpha,5,7aalpha-tetrahydrocyclopenta[c]pyran-4-carboxylate, (1S)-1alpha-(beta-D-Glucopyranosyloxy)-7-(hydroxymethyl)-1,4a alpha,5,7a alpha-tetrahydrocyclopenta[c]pyran-4-carboxylic Acid M
PubChem CID 129316837
IUPAC Name methyl (4aR,7aR)-7-(hydroxymethyl)-1-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate
SMILES COC(=O)C1=COC(C2C1CC=C2CO)OC3C(C(C(C(O3)CO)O)O)O

Specifications

Melting Point 162°C
Color Yellow
Quantity 100 mg
Formula Weight 388.37
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Geniposide