Learn More
CHECKERS™ General-Purpose Non-Lighted Warning Whip with Quick Disconnect Base
Catalog No. 19085960
Change view
Click to view available options
Color:
Orange
Catalog No. Color
19085960 Orange
1 options
Catalog No. 19085960 Supplier CHECKERS™ Supplier No. FS3O
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Non-Lighted Warning Whips include a heavy duty Day Bright™ fluorescent nylon mesh flag
  • Quickly attach or detach from the vehicle
  • Includes Debris Cap to keep unconnected base clean
  • Choose from heavy duty orange, green or yellow flag
  • Whip constructed of premium fiberglass resin
  • Female quick disconnect base mount must be purchased separately. See additional accessories.
  • Quick Disconnect Base has stainless steel components
Specifications
Height (English) 3 ft.
Packaging Quantity 1 Ea.
Color Orange