missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ General Purpose Liquid-in-Glass Total Immersion Thermometers: Blue Organic Fill

With blue organic filling solution. Fisherbrand™ General Purpose Liquid-in-Glass Total Immersion Thermometers are ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing and the petrochemical industry.

Supplier:  Fisherbrand™ B607001120

 View more versions of this product

Catalog No. 13201650


Only null left

Description

Description

  • Traceable to NIST, individually serialized
  • Features blue organic liquid fill against lead-free white back glass
  • Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Tested and inspected by our USA quality team

Specifications

Chemical Identifiers

Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Multilanguage instructions, statement of accuracy
Blue Organic Liquid
Total Immersion
Glass

Specifications

ISO/IEC 17025:2005, A2LA
Total Immersion
7.9 in.
Glass
110°C
-10°C to +110°C
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Multilanguage instructions, statement of accuracy
200 mm
Blue Organic Liquid
-10°C
±1.5°C
Celsius
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions