missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Value Lab Upright Freezers

Meet routine laboratory storage requirements with upright freezers that feature digital controls and excellent temperature stability.

Supplier:  Thermo Scientific™ 04LFEETSA

 View more versions of this product

Catalog No. 04LFEETSA

Only null left

Description

Description

Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature digital controls and excellent temperature stability.

1.6 cu. ft. Model

  • Suitable for -12° to -20°C
  • One fixed interior shelf
  • Key lock
  • UL listed

3.5 cu. ft. Model

  • Suitable for -12° to -20°C
  • Three fixed bins
  • Adjustable thermostat with deep-freeze setting
  • Overtemperature warning with visual alarm
  • Built-in key lock

5 cu. ft. Model

  • Suitable for -12° to -20°C
  • Two fixed shelves
  • Without key lock

Includes: Adjustable shelves or baskets

Specifications

Specifications

General Purpose Freezer
99 L
Adjustable
1 (1 compartment)
Polyurethane Foam
Enameled Steel
cULus
Temperature Display - None
White
Indoor General Storage Of Non-Critical Samples And Reagents
Warranty varies by country, please contact us for details
Interior - No
04LFEETSA
23.5 x 23.63 x.33.5 in. (60 x 61 x 85.1 cm)
Door Lock
NEMA 5-15 P
115 V 60 Hz
160 w
R134a, 100% CFC Free
54.5 kg
45.4 kg
3.5 cu. ft.
-12°C to -20°C
Hinged
Right
ABS Plastic
3
Undercounter
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
Mechanical Dial Thermostat (Outside Front Top Right)
10 Years
Manual
5 in. On Sides, 5 in. At Back, 5 in. On Top
16.5 x 17.75 x 24.25 in. (41.9 x 45.09 x 61.6 cm)
Alarm
1.3 A
115 V
60 Hz
Bottom-mounted
120 lb.
100 lb.
10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions