missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Value Lab Upright Freezers

Catalog No. 02LFEETSA
Change view
Click to view available options
Capacity (English):
1.8 cu. ft.
3.5 cu. ft.
Electrical Requirements:
115 V 60 Hz
Type:
ADA Compliant Freezer
General Purpose Freezer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Electrical Requirements Type
02LFEETSA 1.8 cu. ft. 115 V 60 Hz General Purpose Freezer
04LFEETSA 3.5 cu. ft. 115 V 60 Hz General Purpose Freezer
04LFAETSA 3.5 cu. ft. 115 V 60 Hz ADA Compliant Freezer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02LFEETSA Supplier Thermo Scientific™ Supplier No. 02LFEETSA

Meet routine laboratory storage requirements with upright freezers that feature digital controls and excellent temperature stability.

Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature digital controls and excellent temperature stability.

1.8 cu. ft. Model

  • Suitable for -12° to -24°C
  • One fixed interior shelf
  • Key lock
  • UL listed

3.5 cu. ft. Model

  • Suitable for -12° to -20°C
  • Three fixed bins
  • Adjustable thermostat with deep-freeze setting
  • Overtemperature warning with visual alarm
  • Built-in key lock

5 cu. ft. Model

  • Suitable for -12° to -20°C
  • Two fixed shelves
  • Without key lock

Includes: Adjustable shelves or baskets

Warranty

13 Months Parts and Labor

Specifications

Type General Purpose Freezer
Capacity (English) 1.8 cu. ft.
Capacity (Metric) 51 L
Temperature Range -12°C to -24°C
Temperature (Setpoint) Adjustable
Door Style Hinged
No. of Doors 1 (1 compartment)
Hinge Side Right
Insulation Polyurethane Foam
Interior ABS Plastic
Exterior Finish Enameled Steel
Shelves 1 Removable
Certifications/Compliance cCSAus
Style Undercounter
Display Temperature Display - None
Casters No Leveling Feet
Color White
Control Type Mechanical Dial Thermostat (Outside Rear Bottom Left)
For Use With (Application) Indoor General Storage Of Non-Critical Samples And Reagents
Typical Lifetime 10 Years
Warranty Warranty varies by country, please contact us for details
Defrost Manual
Lighting Interior - no
Min. Clearance 4 in. On Sides, 4 in. At Back, 4 in. On Top
Model 02LFEETSA
Dimensions (D x W x H) Interior 13 x 13.38 x 18.63 in. (33.02 x 34 x 47.32 cm)
Dimensions (L x W x H) Exterior 17.75 x 18.5 x 24.5 in. (45.09 x 47 x 62.23 cm)
Monitoring Options Alarm
Access Security Door Lock
Amperage 1.2 A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V 60 Hz
Frequency 60 Hz
Wattage 85 w
Compressor Bottom-mounted
Refrigerant R134a, 100% CFC Free
Shipping Weight (English) 43 lb.
Shipping Weight (Metric) 19.5 kg
Weight (English) 49 lb.
Weight (Metric) 22 kg
Rack Capacity 10
Show More Show Less