missing translation for 'onlineSavingsMsg'
Learn More

Pointe Scientific General Chemistry Reagents

Catalog No. 23666075
Encompass
Change view
Click to view available options
Quantity:
450 mL
Type:
Alkaline Phosphatase Reagent Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23666075 450 mL Alkaline Phosphatase Reagent Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23666075 Supplier Pointe Scientific Supplier No. A7516450

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For quantitative determination of alkaline phosphatase in human serum

HA416-R1 and HA416-R2 are for Hitachi 747.

  • Methods: p-Npp, kinetic and for Hitachi 747
  • Linearity: 0 to 1000U/L
  • Wavelength: 405nm
  • Stability: 7 days at room temp. and 14 days at 2 to 8°C
  • Reaction Temp.: 3°C
  • Expected Values: Adults 35-123 U/L, Children have a higher normal value

Specifications

Certifications/Compliance CE Marked
Type Alkaline Phosphatase Reagent Kit
Formulation R1 and R2 contains AMP Buffer pH 10.45, p-NPP ≤16mM, Magnesium Ions <1mM, activators and preservatives
For Use With (Application) Quantitative determination of Alkaline Phosphatase in Human Serum, in-vitro diagnostic use only
Quantity 450 mL
Form Liquid