Learn More
General Chemistry Model Kit
Catalog No. S65377
Change view
Click to view available options
Product Type:
Model Kit, General Chemistry
Catalog No. Product Type
S65377 Model Kit, General Chemistry
1 options
Catalog No. S65377 Supplier Customer Channels Group (Ccg) Supplier No. GEN1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Illustrate general chemistry principles
This kit enables students to construct models of a wide variety of organic and inorganic substances. It illustrates VSEPR bonding theory, organic molecular structure, and single, double and triple bonds.
Specifications
For Use With (Application) Illustrates general chemistry principles
Includes Black 4-peg Carbon atom centers, Black 3-peg Carbon atom centers, White 1-peg Hydrogen atom centers, Red 2-peg Oxygen atom centers, Red 4-peg Oxygen atom centers, Yellow 4-peg Sulfur atom centers, Blue 4-peg Nitrogen atom centers, Green 4-peg Halogen atom centers, Green 1-peg Halogen atom centers, Light green 1-peg Halogen atom centers, dark green 1-peg Halogen atom centers, purple 5-peg Phosphorous atom centers, Silver 6-peg Sulfur (xenon) atom centers, 1.25 in. PVC, clear bonds (single bonds), 0.75 in. PVC, clear bonds (sigma bonds), background information booklet, model building activity
Product Type Model Kit, General Chemistry