missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneMapper™ ID-X Software v1.6, client installation

Catalog No. A39978
Change view
Click to view available options
Quantity:
1 License
10 License
5 Licenses
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39978 10 License
A39976 1 License
A39977 5 Licenses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A39978 Supplier Applied Biosystems™ Supplier No. A39978

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Quantity 10 License
Product Line GeneMapper
Product Type GeneMapper ID-X Software V1.6 Client Installation
Shipping Condition Room Temperature
Content And Storage Store at room temperature.