Learn More
Genecon™ Kit
Catalog No. S4391710
Change view
Click to view available options
For Use With (Application):
Basics of electricity & magnetism
Catalog No. For Use With (Application)
S4391710 Basics of electricity & magnetism
1 options
Catalog No. S4391710 Supplier Customer Channels Group (Ccg) Supplier No. B2637
Includes
  • Adventures with the Handheld Generator instruction manual
  • Two regular generators with output cords
  • Modified generator with pulley attached
  • Exoergic experiment device
  • Parallel bulb base
  • Electric swing apparatus
  • Capacitor (5.5V, 1.0F), OG=40
  • Electrolysis apparatus (incl. two test tubes/tongs)
  • Magnetic compass, OG=40
  • Electromagnet set
  • Polarity tester (diode)
  • Nichrome coil
  • Four bulbs in sockets with leads
  • Two rubber magnets
  • Six copper and zinc strips
  • Two test leads with alligator clips
  • Mass-on hook
  • Metal-backed thermometer
  • Nichrome wire
  • 12 extra bulbs
  • Magnetic chips, 1 oz. (25g)
More than 24 experiments covering 10 topics.
  • All of the necessary equipment needed to demonstrate the fundamental properties of electricity and magnetism is included in this kit
  • Main component is the Genecon Handheld Generator (two generators are included in the kit)
  • Designed to achieve an output of six volts
  • Polarity can be changed by reversing the direction of rotation
  • Comprehensive manual that features 24 major experiments
Topics and activities that can be taught using this kit include:
  • Energy conversion (mechanical, potential, electrical, chemical, light)
  • Difference between a motor and a generator
  • Mystery circuit boxes
  • Bulbs in series and in parallel
  • Short circuits and fuses
  • Magnetism and electromagnetism
  • Electrolysis and electroplating
  • Making a motor and motor manipulations
  • Energy storage and lead-acid battery
  • Measuring work
Specifications
For Use With (Application) Basics of electricity & magnetism
Includes Adventures with the handheld generator instruction manual, 2 regular generator with output cord, modified generator with pulley attached, exoergic experiment device, parallel bulb base, electric swing apparatus, capacitor (5.5V, 1F), electrolysis apparatus, magnetic compass, electromagnet set, polarity tester (diode), Nichrome coil, four bulb in socket with Lead, two Rubber magnet, six Copper and Zinc strip, two test Lead with alligator clip, mass-on hook, Metal-backed thermometer, Nichrome wire, 12 extra bulb, magnetic chip