missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gemfibrozil Impurity E, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 500904-61-0 |
| Molecular Formula | C18H26O3 |
| Linear Formula | C18H26O3 |
| Quantity | 10 mg |
| Synonym | (EZ)-2,2-Dimethyl-5-[2,5-dimethyl-4-(propene-1-yl)phenoxy]valeric acid |
| Formula Weight | 290.4 |
| Grade | BP |