missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified

Catalog No. a1413301
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1413301 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1413301 Supplier Gibco™ Supplier No. A1413301
Only null left

A soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors. It is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation.

  • Routinely used for attachment and maintenance of human embryonic stem cells (hESC)
  • Human ESCs have specific needs for attachment and maintenance and some lots of basement membrane extract may not provide adequate support
  • Geltrex™ hESC-qualified reduced growth factor basement membrane matrix has been tested for support of hESC growth and pluripotency, therefore minimizing the need to test multiple lots
  • Consistent manufacturing processes help ensure a protein concentration averaging 15 mg/mL with very little variation from lot-to-lot

Specifications

Cell Type Human induced pluripotent stem cells (iPSC), primary epithelial cells, endothelial cells, smooth muscle cells
Classification Reduced Growth Factor
Concentration 12 to 18 mg/mL
Form Frozen
Product Type Basement Membrane Matrix
Serum Level Serum-free
Sterility Sterile
Without Additives No Phenol Red
Product Line Gibco
Quantity 1 mL
Shipping Condition Dry Ice
Endotoxin Level Low
Format Tube(s)
Purity or Quality Grade Cell Culture Grade
Species Mouse
Content And Storage Geltrex™ LDEV-Free hESC-qualified comes as a sterile protein solution in 1 mL vials.

Store at -20° to -80°C.

Geltrex™ Matrix is stable for 18 months from date of manufacture when properly stored.
Culture Type Adherent Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.