missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified

Catalog No. A1413302
Encompass
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1413302 5 mL
A1413301 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1413302 Supplier Gibco™ Supplier No. A1413302
Only null left

A soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors. It is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation.

  • Routinely used for attachment and maintenance of human embryonic stem cells (hESC)
  • Human ESCs have specific needs for attachment and maintenance and some lots of basement membrane extract may not provide adequate support
  • Geltrex™ hESC-qualified reduced growth factor basement membrane matrix has been tested for support of hESC growth and pluripotency, therefore minimizing the need to test multiple lots
  • Consistent manufacturing processes help ensure a protein concentration averaging 15 mg/mL with very little variation from lot-to-lot

Specifications

Cell Type Human induced pluripotent stem cells (iPSC), primary epithelial cells, endothelial cells, smooth muscle cells
Classification Reduced Growth Factor
Concentration 12 to 18 mg/mL
Form Frozen
Product Type Basement Membrane Matrix
Serum Level Serum-free
Sterility Sterile
Without Additives No Phenol Red
Product Line Gibco
Quantity 5 mL
Shipping Condition Dry Ice
Endotoxin Level Low
Format Tube(s)
Purity or Quality Grade Cell Culture Grade
Species Mouse
Content And Storage Store at -20°C.
Culture Type Adherent Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.