missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Geltrex™ Flex LDEV-Free Reduced Growth Factor Basement Membrane Matrix

Catalog No. A4000046701
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4000046701 1 mL
A4000046702 5 mL
A4000046703 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A4000046701 Supplier Gibco™ Supplier No. A4000046701
Only null left

Geltrex Flex LDEV-Free Reduced Growth Factor Basement Membrane Matrix is a soluble basement membrane matrix purified from EHS tumors. Supports culture of epithelial, endothelial, smooth muscle cells, angiogenesis, neurite outgrowth, bile canaliculi formation assays with polarized hepatocytes.

Gibco Geltrex Flex LDEV-Free Reduced Growth Factor Basement Membrane Matrix is a soluble form of basement membrane that is purified from Engelbreth-Holm-Swarm (EHS) tumor cells. The major components of Geltrex Flex matrix include laminin, collagen IV, entactin, and heparan sulfate proteoglycan.

The solution, which gels at 37°C, can be used for promotion and maintenance of a differentiated phenotype in various cell cultures, including primary epithelial cells, primary neurons, and smooth muscle cells. Geltrex Flex matrix has been used in 3D and 2D cell culture, including angiogenesis assays, neurite outgrowth assays, and bile canaliculi formation assays in polarized hepatocytes for ADME applications.

Features of Geltrex Flex LDEV-Free Reduced Growth Factor Basement Membrane Matrix

  • Each lot tested for the absence of lactose dehydrogenase elevating virus (LDEV), mycoplasma, and a broad panel of viruses and microorganisms
  • Lots tested for endotoxin, maintenance of gel stability, and HUVEC tube formation
  • Growth factor–reduced to improve lot-to-lot consistency
  • Formulated without phenol red to minimize potential for estrogen-like effects
  • Provided in recyclable glass vials
  • 1-mL, 5-mL, and 10-mL sizes for maximum flexibility. Use only what you need. Reduce need for extensive aliquoting and unwanted freeze-thaws.

Gain reliability and consistency with hESC/iPSC-qualified and organoid-qualified Geltrex Flex matrix

For culturing hESCs or iPSCs, it is recommended to use Geltrex Flex LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix. hESC-qualified Geltrex Flex matrix provides all the benefits of standard Geltrex Flex matrix, with added quality controls for each lot. Each lot is tested with iPSCs and H9 hESCs for at least five passages to help ensure reliability and consistency in hESC/iPSC studies.

For culturing tumoroids and other organoids, Geltrex Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix is recommended. Organoid-qualified Geltrex Flex matrix provides all the benefits of standard Geltrex Flex matrix, with added quality controls for each lot. Each lot is tested and supports culture of mouse intestinal organoids, mouse liver ductal organoids, and mouse airway organoids, as well as expansion of mouse small intestinal organoids for more than five passages with typical organoid budding morphology.

Specifications

Cell Type Variety of cultured cells, including HUVEC, primary epithelial cells, neurons, endothelial cells, smooth muscle cells, hepatocytes.
Classification Reduced Growth Factor
Concentration 9 to 13 mg/mL
Form Frozen
Product Type Basement Membrane Matrix
Serum Level Serum-free
Sterility Sterile
Without Additives No Phenol Red
Quantity 1 mL
Shipping Condition Dry Ice
Endotoxin Level ≤ 4.5 EU/mL
Purity or Quality Grade Cell Culture Grade
Species Mouse
Culture Type Adherent Cell Culture, Mammalian Cell Culture, Neuronal Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.