missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ GelRed™ Nucleic Acid Stain 3X Water

Catalog No. sct121
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT121 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT121 Supplier MilliporeSigma™ Supplier No. SCT121
Only null left

GelRed™ is a fluorescent nucleic acid stain designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA, ssDNA or RNA in agarose gels or polyacrylamide gels.

  • GelRed™ is a sensitive, stable and environmentally safe fluorescent nucleic acid dye designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA, ssDNA or RNA in agarose gels or polyacrylamide gels
  • GelRed™ and EtBr have virtually the same spectra, so you can directly replace EtBr with GelRed™ without changing your existing imaging system
  • More sensitive than EtBr
  • The dye is noncytotoxic and non-mutagenic at concentrations well above the working concentrations used in gel staining
  • GelRed™ successfully passed environmental safety tests in compliance with CCR Title 22 Hazardous Waste Characterization, under which GelRed™ is not classified as hazardous waste
  • Absorbance: Standard Transilluminator (302 or 312 nm)
  • Emission: Ethidium Bromide, SYBR or GelStar Filter

Specifications

Description Absorbance: Standard Transilluminator (302 or 312 nm); Emission: Ethidium Bromide, SYBR or GelStar Filter
Detection Fluorescence
Form Liquid
Quantity 4 L
Storage Requirements Store at room temperature, protected from light