missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GelCode™ Blue Stain Reagent

Catalog No. p-4530041
Change view
Click to view available options
Quantity:
500 mL
3.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI24590 500 mL
PI24592 3.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI24590 Supplier Thermo Scientific™ Supplier No. LSG24590
Only null left

GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection.

GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection. This unique reagent stains only protein and allows bands to be viewed directly in the gel during the one-hour gel-staining process.

GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection.

Features include:

  • No methanol-acetic acid destaining required
  • Fast, one-step, one-hour staining, ready-to-use stain reagent; band development visible directly in the staining tray; no background from overnight staining; flexible fixing and washing protocol; no gel shrinkage; stained gel can be dried
  • Water wash increases staining sensitivity; optional step provides crystal-clear gel background and makes weakly stained bands more visible

GelCode Blue Stain Reagent stains only protein and allows bands to be viewed directly in the gel during the one-hour gel-staining process. Because the destaining step does not fix the protein, GelCode Blue Stain is compatible with mass spectrometry analysis and N-terminal sequence analysis.

Specifications

Content And Storage Store at 4°C.
Detection Location In-Blot Detection, In-Gel Detection
Color Blue
Detection Method Colorimetric
Label or Dye Coomassie
Quantity 500 mL
Target Molecule Protein
Product Line GelCode
Product Type Protein Gel Stain