missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GelCode™ Blue Safe Protein Stain

Catalog No. p-4530026
Change view
Click to view available options
Quantity:
1 L
3.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI24594 1 L
PI24596 3.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI24594 Supplier Thermo Scientific™ Supplier No. LSG24594
Only null left

GelCode Blue Safe Protein Stain is a fast and sensitive coomassie-dye reagent that provides one-step ultrasensitive detection of proteins in polyacrylamide gels and membranes.

GelCode Blue Safe Protein Stain is a fast and sensitive coomassie-dye reagent that provides one-step ultrasensitive detection of proteins in polyacrylamide gels and membranes. This protein-specific stain allows bands to be viewed directly on a gel or membrane during staining and is compatible with mass spectrometry applications. GelCode Blue Safe Protein Stain is non-corrosive, non-flammable, and is classified as non-hazardous under the U.S. Department of Transportation (DOT) shipping requirements.

GelCode Blue Safe Protein Stain is a fast and sensitive coomassie-dye reagent that provides one-step ultrasensitive detection of proteins in polyacrylamide gels and is formulated to avoid hazardous shipping.

Features include:

  • Sensitive—detect down to 9 ng of protein per band in 15 minutes
  • Safe—non-corrosive to skin, non-flammable, and safe to ship and store until activated
  • Minimal additional reagents—water washes only; no acid-fixative or methanol destain solutions required
  • Compatible—suitable for staining 1D and 2D polyacrylamide gels and nitrocellulose and PVDF membranes (multiple protocols provided in the instructions)
  • Multiple applications—qualitative visualization, quantitative densitometry, and downstream excision and analysis by mass spectrometry
  • Easy to use—add and dissolve activator crystals before first use, then simply add reagent to stain gels
  • Stable—store non-activated or activated stain at room temperature for up to one year

Because the supplied activator crystals are packaged and shipped dry in a separate bottle, GelCode Blue Safe Protein Stain is classified as nonhazardous under the U.S. Department of Transportation (DOT) shipping requirements. For proper protein staining, the activator crystals must be dissolved in the staining reagent before use. Add the entire contents of the activator crystals bottle into the GelCode Blue Safe Protein Stain bottle. Dissolve the crystals by mixing end-over-end for one minute, then allow it to remain at room temperature for five minutes before use. Store the activated stain at room temperature.

Specifications

Content And Storage Store at room temperature.
Detection Location In-Blot Detection, In-Gel Detection
Color Blue
Detection Method Colorimetric
Label or Dye Coomassie
Quantity 1 L
Target Molecule Protein
Product Line GelCode
Product Type Protein Stain